siRNA Details
| Gene Symbol: Stc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022230.2 | NP_071566.1 | 3 | 388 | ||
| GenBank mRNA | AB030707.1 | - | 3 | 388 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stc2| Gene Symbol: Stc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022230.2 | NP_071566.1 | 3 | 388 | ||
| GenBank mRNA | AB030707.1 | - | 3 | 388 | ||