siRNA Details
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022230.2 | NP_071566.1 | 3, 4 | 528 | ||
GenBank mRNA | AB030707.1 | - | 3, 4 | 528 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022230.2 | NP_071566.1 | 3, 4 | 528 | ||
GenBank mRNA | AB030707.1 | - | 3, 4 | 528 |