siRNA Details
| Gene Symbol: Cyb5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022245.1 | NP_071581.1 | 5 | 537 | ||
| GenBank mRNA | BC086945.1 | - | 5 | 531 | 7322141 | |
| D13205.1 | - | 5 | 537 | |||
| FQ209861.1 | - | 5 | 528 | |||
| FQ210790.1 | - | 5 | 525 | |||
| FQ211029.1 | - | 3 | 213 | |||
| FQ224354.1 | - | 5 | 528 | |||

Cyb5a
Genomic Map