siRNA Details
| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022251.2 | NP_071587.2 | 14 | 2501 | ||
| GenBank mRNA | AF146518.1 | - | 13 | 1783 | ||
| AF214568.1 | - | 14 | 2318 | |||
| BC066663.1 | - | 14 | 2323 | 5623429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Enpep| Gene Symbol: Enpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022251.2 | NP_071587.2 | 14 | 2501 | ||
| GenBank mRNA | AF146518.1 | - | 13 | 1783 | ||
| AF214568.1 | - | 14 | 2318 | |||
| BC066663.1 | - | 14 | 2323 | 5623429 | ||