siRNA Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022260.3 | NP_071596.1 | 1 | 98 | ||
| XM_008760546.1 | XP_008758768.1 | 2 | 691 | |||
| GenBank mRNA | AF072124.1 | - | 1 | 99 | ||
| BC070936.1 | - | 1 | 99 | 7099111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casp7| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022260.3 | NP_071596.1 | 1 | 98 | ||
| XM_008760546.1 | XP_008758768.1 | 2 | 691 | |||
| GenBank mRNA | AF072124.1 | - | 1 | 99 | ||
| BC070936.1 | - | 1 | 99 | 7099111 | ||