siRNA Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022264.1 | NP_071600.1 | 2 | 366 | ||
| XM_006250909.2 | XP_006250971.1 | 2 | 400 | |||
| GenBank mRNA | D12524.1 | - | 2 | 366 | ||
| EU247827.1 | - | 2 | 350 | |||
| EU247828.1 | - | 2 | 350 | |||

Kit
Genomic Map