siRNA Details
| Gene Symbol: Qdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022390.1 | NP_071785.1 | 3 | 268 | ||
| GenBank mRNA | BC072536.1 | - | 3 | 281 | 7103182 | |
| FQ213296.1 | - | 3 | 247 | |||
| FQ213843.1 | - | 3 | 298 | |||
| J03481.1 | - | 3 | 268 | |||

Qdpr
Genomic Map