siRNA Details
| Gene Symbol: RAB35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167606.1 | NP_001161078.1 | 5 | 2039 | ||
| NM_006861.6 | NP_006852.1 | 6 | 2164 | |||
| XM_011537779.1 | XP_011536081.1 | 6 | 2101 | |||
| XM_017018723.1 | XP_016874212.1 | 5 | 1941 | |||
| GenBank mRNA | AK001309.2 | - | 1 | 966 | ||
| BC015931.2 | - | 6 | 2079 | 3907209 | ||
| BC017753.1 | - | 1 | 665 | |||

RAB35
Genomic Map