siRNA Details
| Gene Symbol: LILRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF025529.1 | - | 8 | 1244 | ||
| AF025530.1 | - | 10 | 1856 | |||
| AK290919.1 | - | 10 | 1872 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LILRA1| Gene Symbol: LILRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF025529.1 | - | 8 | 1244 | ||
| AF025530.1 | - | 10 | 1856 | |||
| AK290919.1 | - | 10 | 1872 | |||