siRNA Details
| Gene Symbol: Ppap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022538.2 | NP_071983.2 | 5 | 786 | ||
| XM_006231949.2 | XP_006232011.1 | 5 | 1154 | |||
| GenBank mRNA | AF503609.1 | - | 5 | 692 | ||
| AF503610.1 | - | 4 | 537 | |||
| BC061815.1 | - | 5 | 786 | 5598693 | ||
| FQ229687.1 | - | 3 | 381 | |||
| JQ013728.1 | - | 5 | 724 | |||
| U90556.1 | - | 5 | 701 | |||

Ppap2a
Genomic Map