siRNA Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022597.2 | NP_072119.2 | 3 | 311 | ||
| GenBank mRNA | BC072490.1 | - | 3 | 274 | 7097817 | |
| X82396.1 | - | 3 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsb| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022597.2 | NP_072119.2 | 3 | 311 | ||
| GenBank mRNA | BC072490.1 | - | 3 | 274 | 7097817 | |
| X82396.1 | - | 3 | 240 | |||