siRNA Details
| Gene Symbol: Inhbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022614.2 | NP_072136.1 | 2 | 538 | ||
| GenBank mRNA | AF140031.1 | - | 2 | 445 | ||
| BC089799.1 | - | 2 | 538 | 7321367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inhbc| Gene Symbol: Inhbc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022614.2 | NP_072136.1 | 2 | 538 | ||
| GenBank mRNA | AF140031.1 | - | 2 | 445 | ||
| BC089799.1 | - | 2 | 538 | 7321367 | ||