siRNA Details
| Gene Symbol: Nphs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022628.1 | NP_072150.1 | 30 | 5461 | ||
| GenBank mRNA | AF161715.1 | - | 30 | 5461 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nphs1| Gene Symbol: Nphs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022628.1 | NP_072150.1 | 30 | 5461 | ||
| GenBank mRNA | AF161715.1 | - | 30 | 5461 | ||