siRNA Details
| Gene Symbol: Zic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022677.1 | NP_073168.1 | 1 | 680 | ||
| XM_008766454.1 | XP_008764676.1 | 2 | 1048 | |||
| GenBank mRNA | AF221839.1 | - | 1 | 680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zic1| Gene Symbol: Zic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022677.1 | NP_073168.1 | 1 | 680 | ||
| XM_008766454.1 | XP_008764676.1 | 2 | 1048 | |||
| GenBank mRNA | AF221839.1 | - | 1 | 680 | ||