siRNA Details
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022866.2 | NP_074057.2 | 7 | 999 | ||
GenBank mRNA | AF080451.1 | - | 7 | 976 | ||
AF081825.1 | - | 7 | 1001 | |||
BC129094.1 | - | 7 | 999 | 8365494 |
