siRNA Details
| Gene Symbol: Bche | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022942.1 | NP_075231.1 | 2 | 296 | ||
| XM_008761112.1 | XP_008759334.1 | 1 | 373 | |||
| GenBank mRNA | AF244349.1 | - | 2 | 296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bche| Gene Symbol: Bche | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022942.1 | NP_075231.1 | 2 | 296 | ||
| XM_008761112.1 | XP_008759334.1 | 1 | 373 | |||
| GenBank mRNA | AF244349.1 | - | 2 | 296 | ||