siRNA Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023963.1 | NP_076453.1 | 3 | 1222 | ||
| XM_008768999.1 | XP_008767221.1 | 3 | 1113 | |||
| GenBank mRNA | AJ278466.1 | - | 3 | 1222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdx2| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023963.1 | NP_076453.1 | 3 | 1222 | ||
| XM_008768999.1 | XP_008767221.1 | 3 | 1113 | |||
| GenBank mRNA | AJ278466.1 | - | 3 | 1222 | ||