siRNA Details
| Gene Symbol: Prx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023976.2 | NP_076466.2 | 6 | 612 | ||
| XM_006228586.2 | XP_006228648.1 | 6 | 705 | |||
| XM_008759196.1 | XP_008757418.1 | 5 | 806 | |||
| GenBank mRNA | Z29649.2 | - | 6 | 612 | ||

Prx
Genomic Map