siRNA Details
| Gene Symbol: Birc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023987.3 | NP_076477.3 | 3 | 878 | ||
| XM_006242499.2 | XP_006242561.1 | 2 | 2841 | |||
| XM_006242500.2 | XP_006242562.1 | 3 | 1611 | |||
| XM_006242502.2 | XP_006242564.1 | 2 | 1110 | |||
| XM_006242503.2 | XP_006242565.1 | 2 | 1069 | |||
| GenBank mRNA | AF183430.1 | - | 1 | 646 | ||
| BC083555.1 | - | 3 | 878 | 7112819 | ||

Birc3
Genomic Map