siRNA Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024146.1 | NP_077060.1 | 1, 2 | 85 | ||
| XM_006253323.2 | XP_006253385.1 | 1, 2 | 93 | |||
| XM_006253324.2 | XP_006253386.1 | 2, 3 | 306 | |||
| GenBank mRNA | D12498.1 | - | 1, 2 | 85 | ||

Fgfr1
Genomic Map