siRNA Details
| Gene Symbol: Arf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024151.1 | NP_077065.1 | 6 | 859 | ||
| GenBank mRNA | BC063167.1 | - | 6 | 817 | 6921397 | |
| FQ213804.1 | - | 6 | 858 | |||
| L12383.1 | - | 6 | 859 | |||
| M86705.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arf4| Gene Symbol: Arf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024151.1 | NP_077065.1 | 6 | 859 | ||
| GenBank mRNA | BC063167.1 | - | 6 | 817 | 6921397 | |
| FQ213804.1 | - | 6 | 858 | |||
| L12383.1 | - | 6 | 859 | |||
| M86705.1 | - | 1 | 100 | |||