siRNA Details
| Gene Symbol: Galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024373.1 | NP_077349.1 | 4 | 629 | ||
| XM_006254471.2 | XP_006254533.1 | 5 | 2050 | |||
| GenBank mRNA | U35890.1 | - | 4 | 629 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt1| Gene Symbol: Galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024373.1 | NP_077349.1 | 4 | 629 | ||
| XM_006254471.2 | XP_006254533.1 | 5 | 2050 | |||
| GenBank mRNA | U35890.1 | - | 4 | 629 | ||