siRNA Details
| Gene Symbol: Hhex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024385.1 | NP_077361.1 | 2 | 468 | ||
| GenBank mRNA | BC088135.1 | - | 2 | 473 | 7366155 | |
| D86383.1 | - | 2 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhex| Gene Symbol: Hhex | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024385.1 | NP_077361.1 | 2 | 468 | ||
| GenBank mRNA | BC088135.1 | - | 2 | 473 | 7366155 | |
| D86383.1 | - | 2 | 468 | |||