siRNA Details
| Gene Symbol: Hmgcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024386.1 | NP_077362.1 | 4 | 346 | ||
| GenBank mRNA | BC061797.1 | - | 4 | 348 | 5598033 | |
| FQ209392.1 | - | 4 | 344 | |||
| FQ209616.1 | - | 4 | 350 | |||
| FQ234382.1 | - | 4 | 344 | |||
| Y10054.1 | - | 4 | 346 | |||

Hmgcl
Genomic Map