siRNA Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024392.2 | NP_077368.2 | 2, 3 | 157 | ||
| GenBank mRNA | BC072472.1 | - | 2, 3 | 147 | 7100430 | |
| S83279.1 | - | 2, 3 | 129 | |||
| U37486.1 | - | 2, 3 | 157 | |||
| X94978.1 | - | 2, 3 | 132 | |||

Hsd17b4
Genomic Map