siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030844.2 | NP_110471.2 | 9 | 1079 | ||
| XM_006236092.2 | XP_006236154.1 | 9 | 1175 | |||
| XM_006236093.2 | XP_006236155.1 | 9 | 1397 | |||
| XM_006236094.2 | XP_006236156.1 | 8 | 1025 | |||
| XM_006236095.2 | XP_006236157.1 | 8 | 1011 | |||
| XM_008762728.1 | XP_008760950.1 | 9 | 1175 | |||
| GenBank mRNA | BC063810.1 | - | 9 | 1098 | ||
| BC078812.1 | - | 9 | 1079 | 7112043 | ||
| L20900.1 | - | 9 | 1285 | |||

Ica1
Genomic Map