siRNA Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030846.2 | NP_110473.2 | 5 | 585 | ||
| GenBank mRNA | BC091144.1 | - | 4 | 529 | 7382476 | |
| D49846.1 | - | 5 | 700 | |||
| D49848.1 | - | 4 | 400 | |||
| FQ220840.1 | - | 5 | 651 | |||
| X62853.1 | - | 4 | 435 | |||
| Gene Symbol: LOC103690155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768457.1 | XP_008766679.1 | 4 | 476 | ||

Grb2
Genomic Map