siRNA Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030846.2 | NP_110473.2 | 6 | 1165 | ||
| GenBank mRNA | BC091144.1 | - | 5 | 1109 | 7382476 | |
| D49846.1 | - | 6 | 1238 | |||
| D49848.1 | - | 5 | 980 | |||
| X62853.1 | - | 5 | 1015 | |||

Grb2
Genomic Map