siRNA Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030850.1 | NP_110477.1 | 8 | 1686 | ||
| GenBank mRNA | AF038870.1 | - | 8 | 1686 | ||
| BC078811.1 | - | 8 | 1673 | 7130736 | ||
| FQ218356.1 | - | 3 | 1011 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhmt| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030850.1 | NP_110477.1 | 8 | 1686 | ||
| GenBank mRNA | AF038870.1 | - | 8 | 1686 | ||
| BC078811.1 | - | 8 | 1673 | 7130736 | ||
| FQ218356.1 | - | 3 | 1011 | |||