siRNA Details
| Gene Symbol: Marcksl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030862.2 | NP_110489.1 | 1, 2 | 277 | ||
| GenBank mRNA | AJ301677.1 | - | 1, 2 | 170 | ||
| BC081789.1 | - | 1, 2 | 277 | 7134056 | ||
| Gene Symbol: LOC103689966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759211.1 | XP_008757433.1 | 1 | 311 | ||

Marcksl1
Genomic Map