siRNA Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030867.2 | NP_110494.2 | 3 | 864 | ||
| GenBank mRNA | AF246634.1 | - | 3 | 687 | ||
| BC085729.1 | - | 3 | 864 | 7114624 | ||
| FQ235103.1 | - | 3 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfkbib| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030867.2 | NP_110494.2 | 3 | 864 | ||
| GenBank mRNA | AF246634.1 | - | 3 | 687 | ||
| BC085729.1 | - | 3 | 864 | 7114624 | ||
| FQ235103.1 | - | 3 | 810 | |||