siRNA Details
| Gene Symbol: Nov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030868.2 | NP_110495.1 | 3 | 401 | ||
| GenBank mRNA | AF171936.1 | - | 3 | 388 | ||
| BC072548.1 | - | 3 | 401 | 7105981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nov| Gene Symbol: Nov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030868.2 | NP_110495.1 | 3 | 401 | ||
| GenBank mRNA | AF171936.1 | - | 3 | 388 | ||
| BC072548.1 | - | 3 | 401 | 7105981 | ||