siRNA Details
| Gene Symbol: Pfn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030873.1 | NP_110500.1 | 796 | |||
| XM_006232390.2 | XP_006232452.1 | 3 | 922 | |||
| GenBank mRNA | AY004289.1 | - | 796 | |||
| BC087013.1 | - | 800 | ||||
| FQ215807.1 | - | 1 | 173 | |||
| Gene Symbol: LOC100909840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749282.2 | XP_003749330.1 | 3 | 838 | ||

Pfn2
Genomic Map