siRNA Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030991.3 | NP_112253.1 | 373 | |||
| GenBank mRNA | AB003992.1 | - | 164 | |||
| AF245227.1 | - | 372 | ||||
| BC087699.1 | - | 328 | 7314318 | |||
| FQ212040.1 | - | 344 | ||||
| FQ213289.1 | - | 4 | 341 | |||
| FQ213923.1 | - | 344 | ||||
| FQ214162.1 | - | 373 | ||||
| U56262.1 | - | 137 | ||||

Snap25
Genomic Map