siRNA Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270575.1 | NP_001257504.1 | 7 | 1706 | ||
| NM_001270576.1 | NP_001257505.1 | 7 | 1618 | |||
| NM_030991.3 | NP_112253.1 | 7 | 1706 | |||
| GenBank mRNA | AF245227.1 | - | 7 | 1706 | ||
| BC087699.1 | - | 7 | 1661 | 7314318 | ||
| FQ212040.1 | - | 8 | 962 | |||

Snap25
Genomic Map