siRNA Details
| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031027.1 | NP_112289.1 | 5 | 403 | ||
| GenBank mRNA | D85035.1 | - | 5 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpyd| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031027.1 | NP_112289.1 | 5 | 403 | ||
| GenBank mRNA | D85035.1 | - | 5 | 403 | ||