siRNA Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031029.1 | NP_112291.1 | 3 | 265 | ||
| GenBank mRNA | FQ222085.1 | - | 4 | 432 | ||
| U95368.1 | - | 3 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrp| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031029.1 | NP_112291.1 | 3 | 265 | ||
| GenBank mRNA | FQ222085.1 | - | 4 | 432 | ||
| U95368.1 | - | 3 | 265 | |||