siRNA Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031029.1 | NP_112291.1 | 4 | 303 | ||
| XM_008767594.1 | XP_008765816.1 | 5 | 889 | |||
| XM_008767595.1 | XP_008765817.1 | 5 | 895 | |||
| GenBank mRNA | FQ222085.1 | - | 5 | 470 | ||
| U95368.1 | - | 4 | 303 | |||

Gabrp
Genomic Map