siRNA Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031048.1 | NP_112310.1 | 9 | 1262 | ||
| XM_006232027.2 | XP_006232089.1 | 9 | 1436 | |||
| XM_006232029.2 | XP_006232091.1 | 9 | 2097 | |||
| XM_008760796.1 | XP_008759018.1 | 9 | 2379 | |||
| GenBank mRNA | D86345.1 | - | 9 | 1262 | ||

Lifr
Genomic Map