siRNA Details
| Gene Symbol: Lss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031049.1 | NP_112311.1 | 3 | 260 | ||
| GenBank mRNA | D45252.1 | - | 3 | 284 | ||
| U31352.1 | - | 3 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lss| Gene Symbol: Lss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031049.1 | NP_112311.1 | 3 | 260 | ||
| GenBank mRNA | D45252.1 | - | 3 | 284 | ||
| U31352.1 | - | 3 | 260 | |||