siRNA Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031052.1 | NP_112314.1 | 18 | 2108 | ||
| XM_008770779.1 | XP_008769001.1 | 17 | 2186 | |||
| GenBank mRNA | M96633.1 | - | 18 | 2108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mipep| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031052.1 | NP_112314.1 | 18 | 2108 | ||
| XM_008770779.1 | XP_008769001.1 | 17 | 2186 | |||
| GenBank mRNA | M96633.1 | - | 18 | 2108 | ||