siRNA Details
| Gene Symbol: Mmp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031055.1 | NP_112317.1 | 2 | 274 | ||
| GenBank mRNA | U24441.1 | - | 2 | 274 | ||
| U36476.1 | - | 2 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp9| Gene Symbol: Mmp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031055.1 | NP_112317.1 | 2 | 274 | ||
| GenBank mRNA | U24441.1 | - | 2 | 274 | ||
| U36476.1 | - | 2 | 274 | |||