siRNA Details
| Gene Symbol: Prkch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031085.2 | NP_112347.1 | 2, 3 | 611 | ||
| XM_008764720.1 | XP_008762942.1 | 2, 3 | 493 | |||
| GenBank mRNA | BC061763.1 | - | 3, 4 | 942 | ||
| BC081782.1 | - | 2, 3 | 611 | 7123785 | ||
| X68400.1 | - | 2, 3 | 452 | |||

Prkch
Genomic Map