siRNA Details
Gene Symbol: Doc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031142.1 | NP_112404.1 | 1 | 256 | ||
XM_008768117.1 | XP_008766339.1 | 1 | 323 | |||
GenBank mRNA | U70778.2 | - | 1 | 256 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Doc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031142.1 | NP_112404.1 | 1 | 256 | ||
XM_008768117.1 | XP_008766339.1 | 1 | 323 | |||
GenBank mRNA | U70778.2 | - | 1 | 256 |