siRNA Details
| Gene Symbol: Cirbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031147.2 | NP_112409.2 | 6, 7 | 674 | ||
| XM_006241014.2 | XP_006241076.1 | 6, 7 | 679 | |||
| XM_006241015.2 | XP_006241077.2 | 6, 7 | 790 | |||
| GenBank mRNA | AB000362.1 | - | 5, 6 | 523 | ||
| BC069219.1 | - | 6, 7 | 674 | 6888887 | ||

Cirbp
Genomic Map