siRNA Details
| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031242.2 | NP_112521.2 | 1, 2 | 171 | ||
| GenBank mRNA | AB009999.1 | - | 116 | |||
| BC127492.1 | - | 1, 2 | 171 | 8373547 | ||
| FQ212493.1 | - | 146 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173370, AF533367, BC055292.

Cds1
Genomic Map