siRNA Details
| Gene Symbol: Celsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031320.1 | NP_112610.1 | 1 | 657 | ||
| XM_006243860.2 | XP_006243922.1 | 1 | 394 | |||
| XM_006243861.2 | XP_006243923.1 | 1 | 394 | |||
| GenBank mRNA | AB011528.1 | - | 1 | 657 | ||
More Information
Set Membership: |
|

Celsr3
Genomic Map