siRNA Details
| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031325.1 | NP_112615.1 | 3 | 362 | ||
| GenBank mRNA | AB013732.1 | - | 3 | 362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugdh| Gene Symbol: Ugdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031325.1 | NP_112615.1 | 3 | 362 | ||
| GenBank mRNA | AB013732.1 | - | 3 | 362 | ||