siRNA Details
| Gene Symbol: Cyr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031327.2 | NP_112617.2 | 4, 5 | 1020 | ||
| GenBank mRNA | AB015877.1 | - | 4, 5 | 885 | ||
| AF218568.1 | - | 4, 5 | 1020 | |||
| BC081689.1 | - | 4, 5 | 1000 | 7122117 | ||

Cyr61
Genomic Map