siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 4, 5 | 709 | ||
| XM_008760243.1 | XP_008758465.1 | 3, 4 | 524 | |||
| GenBank mRNA | AB017446.1 | - | 4, 5 | 709 | ||
| BC081777.1 | - | 4, 5 | 707 | 7109603 | ||
| FQ209755.1 | - | 721 | ||||

Slc22a8
Genomic Map